accessibility menu, dialog, popup

UserName

CAS RN 2924-15-4

IUPAC Name: hydrogen (2-fluorophenyl)hydrazine chloride
Synonyms: hydrogen (2-fluorophenyl)hydrazine chloride
Molecular Weight (g/mol): 162.59
Molecular Formula: C6H8ClFN2
InChi Key: VFZYLSYYMHFPSY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=CC=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)