missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylhydrazine Hydrochloride 97.0+%, TCI America™

Catalog No. F03685G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03685G 5 g
F03681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03685G Supplier TCI America Supplier No. F03685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2924-15-4
Molecular Formula C6H8ClFN2
Molecular Weight (g/mol) 162.592
MDL Number MFCD00012927
InChI Key VFZYLSYYMHFPSY-UHFFFAOYSA-N
Synonym 2-fluorophenylhydrazine hydrochloride, 2-fluorophenyl hydrazine hydrochloride, 2-fluorophenylhydrazine hcl, 2-fluorophenylhydrazinehydrochloride, o-fluorophenylhydrazine hydrochloride, hydrazine, 2-fluorophenyl-, monohydrochloride, 1-2-fluorophenyl hydrazine hydrochloride, pubchem3347, c6h7fn2.hcl, acmc-209h7m
PubChem CID 2723910
IUPAC Name (2-fluorophenyl)hydrazine;hydrochloride
SMILES C1=CC=C(C(=C1)NN)F.Cl

Specifications

Quantity 5 g
Formula Weight 162.59
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluorophenylhydrazine Hydrochloride