accessibility menu, dialog, popup

UserName

CAS RN 2916-31-6

IUPAC Name: 2,2-dimethyl-1,3-dioxolane
Synonyms: 1,3-dioxolane, 2,2-dimethyl-
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: SIJBDWPVNAYVGY-UHFFFAOYSA-N
SMILES: CC1(C)OCCO1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)