missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-dioxolane, 98+%

Catalog No. AAL0048214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0048214 25 g
AAL0048206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0048214 Supplier Thermo Scientific Chemicals Supplier No. L0048214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2916-31-6 | C5H10O2 | 102.13 g/mol

2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.

Solubility
Insoluble in water.

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 2916-31-6
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00014106
InChI Key SIJBDWPVNAYVGY-UHFFFAOYSA-N
PubChem CID 76209
IUPAC Name 2,2-dimethyl-1,3-dioxolane
SMILES CC1(C)OCCO1

Specifications

Density 0.946
Boiling Point 91°C to 93°C
Flash Point 6°C (42°F)
Refractive Index 1.398
Quantity 25 g
UN Number UN1993
Beilstein 102775
Synonym 1,3-dioxolane, 2,2-dimethyl, 2,2-dimethyl-1,3-dioxolan, 1,3-dioxolane, dimethyl, acetone ethylene ketal, acetone ethylene acetal, acetone, cyclic ethylene acetal, 1,3-dioxolane,2,2-dimethyl, sijbdwpvnayvgy-uhfffaoysa, 2,2-dimethyl-1,3-dioxolane
Solubility Information Insoluble in water.
Formula Weight 102.13
Percent Purity ≥98%
Chemical Name or Material 2,2-Dimethyl-1,3-dioxolane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 220-842-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only