accessibility menu, dialog, popup

UserName

ClOOCH2

CAS RN 2916-14-5

IUPAC Name: prop-2-en-1-yl 2-chloroacetate
Synonyms: prop-2-en-1-yl 2-chloroacetate
Molecular Weight (g/mol): 134.56
Molecular Formula: C5H7ClO2
InChi Key: VMBJJCDVORDOCF-UHFFFAOYSA-N
SMILES: ClCC(=O)OCC=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)