missing translation for 'onlineSavingsMsg'
Learn More

Allyl Chloroacetate 98.0+%, TCI America™

Catalog No. C0090500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0090500G 500 g
C009025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0090500G Supplier TCI America Supplier No. C0090500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2916-14-5
Molecular Formula C5H7ClO2
Molecular Weight (g/mol) 134.559
MDL Number MFCD00058936
InChI Key VMBJJCDVORDOCF-UHFFFAOYSA-N
Synonym allyl chloroacetate, allyl 2-chloroacetate, chloroacetic acid allyl ester, acetic acid, chloro-, 2-propenyl ester, prop-2-en-1-yl 2-chloroacetate, acetic acid, chloro-, allyl ester, allyl monochloroacetate, acmc-1cffa, chloroacetic acid,2-propenyl ester
PubChem CID 76206
IUPAC Name prop-2-enyl 2-chloroacetate
SMILES C=CCOC(=O)CCl

Specifications

Color Yellow
Boiling Point 164°C
Quantity 500 g
Formula Weight 134.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Chloroacetate