accessibility menu, dialog, popup

UserName

CAS RN 288-47-1

IUPAC Name: 1,3-thiazole
Synonyms: thiazole
Molecular Weight (g/mol): 85.12
Molecular Formula: C3H3NS
InChi Key: FZWLAAWBMGSTSO-UHFFFAOYSA-N
SMILES: S1C=CN=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (7)
  • (2)
  • (3)
  • (2)