missing translation for 'onlineSavingsMsg'
Learn More

Thiazole 98.0+%, TCI America™

Catalog No. T01855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01855G 5 g
T018525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T01855G Supplier TCI America Supplier No. T01855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 288-47-1
Molecular Formula C3H3NS
Molecular Weight (g/mol) 85.12
MDL Number MFCD00005315
InChI Key FZWLAAWBMGSTSO-UHFFFAOYSA-N
Synonym thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium
PubChem CID 9256
ChEBI CHEBI:43732
IUPAC Name 1,3-thiazole
SMILES S1C=CN=C1

Specifications

Melting Point -33°C
Color Yellow
Boiling Point 118°C
Quantity 5 g
UN Number 1993
Formula Weight 85.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Thiazole
RTECSNumber XJ1290000
TSCA Yes