accessibility menu, dialog, popup

UserName

CAS RN 2873-18-9

IUPAC Name: 2-bromo-5-chlorothiophene
Synonyms: 2-bromo-5-chlorothiophene
Molecular Weight (g/mol): 197.47
Molecular Formula: C4H2BrClS
InChi Key: ZFAJPWYXLYGUJU-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Br)S1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)