missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chlorothiophene 95.0+%, TCI America™

Catalog No. B19965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19965G 5 g
B199625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19965G Supplier TCI America Supplier No. B19965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2873-18-9
Molecular Formula C4H2BrClS
Molecular Weight (g/mol) 197.47
MDL Number MFCD00014523
InChI Key ZFAJPWYXLYGUJU-UHFFFAOYSA-N
Synonym 2-chloro-5-bromothiophene, 5-bromo-2-chlorothiophene, 2-chloro-5-bromo thiophene, thiophene, 2-bromo-5-chloro, 2-brom-5-chlorthiophen, pubchem5279, acmc-1cs3j, 2-bromo-5-chloro-thiophene, 5-bromo-2-chloro-thiophene, ksc207m3t
PubChem CID 76133
IUPAC Name 2-bromo-5-chlorothiophene
SMILES ClC1=CC=C(Br)S1

Specifications

Boiling Point 70°C
Quantity 5 g
Formula Weight 197.47
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-chlorothiophene