accessibility menu, dialog, popup

UserName

CAS RN 28721-07-5

IUPAC Name: 9-oxo-2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-2-carboxamide
Synonyms: oxcarbazepine timox desitin brand of oxcarbazepine oxcarbazepine (product) 奥卡西平 oxcarbazepine (substance) trileptal oxcarbazepinum oxcarbazepina novartis brand of oxcarbazepine
Molecular Weight (g/mol): 252.27
Molecular Formula: C15H12N2O2
InChi Key: CTRLABGOLIVAIY-UHFFFAOYSA-N
SMILES: NC(=O)N1C2=CC=CC=C2CC(=O)C2=CC=CC=C12
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)