accessibility menu, dialog, popup

UserName

CAS RN 2870-32-8

IUPAC Name: disodium 5-[2-(4-ethoxyphenyl)diazen-1-yl]-2-[(1Z)-2-{4-[2-(4-ethoxyphenyl)diazen-1-yl]-2-sulfonatophenyl}ethenyl]benzene-1-sulfonate
Synonyms: disodium 5-[2-(4-ethoxyphenyl)diazen-1-yl]-2-[(1Z)-2-{4-[2-(4-ethoxyphenyl)diazen-1-yl]-2-sulfonatophenyl}ethenyl]benzenesulfonate
Molecular Weight (g/mol): 680.66
Molecular Formula: C30H26N4Na2O8S2
InChi Key: YQMJDPHTMKUEHG-RPKDOIGLSA-L
SMILES: [Na+].[Na+].CCOC1=CC=C(C=C1)N=NC1=CC=C(\C=C/C2=CC=C(C=C2S([O-])(=O)=O)N=NC2=CC=C(OCC)C=C2)C(=C1)S([O-])(=O)=O
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)