missing translation for 'onlineSavingsMsg'
Learn More

Chrysophenine, TCI America™

Catalog No. C053025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C053025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C053025G Supplier TCI America Supplier No. C053025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2870-32-8
Molecular Formula C30H26N4Na2O8S2
Molecular Weight (g/mol) 680.66
MDL Number MFCD00007488
InChI Key YQMJDPHTMKUEHG-RPKDOIGLSA-L
Synonym chrysophenine g, chrysophenine j, chrysophenine y, aurophenine o, chrysophenine gp, chrysophenine ns, direct yellow c, benzo yellow lg, erie yellow y, chrysophenine esp
PubChem CID 54603156
IUPAC Name disodium 5-[2-(4-ethoxyphenyl)diazen-1-yl]-2-[(1Z)-2-{4-[2-(4-ethoxyphenyl)diazen-1-yl]-2-sulfonatophenyl}ethenyl]benzene-1-sulfonate
SMILES [Na+].[Na+].CCOC1=CC=C(C=C1)N=NC1=CC=C(\C=C/C2=CC=C(C=C2S([O-])(=O)=O)N=NC2=CC=C(OCC)C=C2)C(=C1)S([O-])(=O)=O

Specifications

Quantity 25 g
Formula Weight 680.66
Physical Form Crystalline Powder
Chemical Name or Material Chrysophenine
EINECSNumber (5)-1182
TSCA Yes