accessibility menu, dialog, popup

UserName

CH3BrBrCH3

CAS RN 286438-45-7

IUPAC Name: 2,7-dibromo-9,9-didodecyl-9H-fluorene
Synonyms: 2,7-dibromo-9,9-didodecylfluorene
Molecular Weight (g/mol): 660.66
Molecular Formula: C37H56Br2
InChi Key: KFOUJVGPGBSIFB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1(CCCCCCCCCCCC)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)