accessibility menu, dialog, popup

UserName

H3CNCH3BOHHO

CAS RN 28611-39-4

IUPAC Name: [4-(dimethylamino)phenyl]boronic acid
Synonyms: 4-(dimethylamino)phenylboronic acid
Molecular Weight (g/mol): 165
Molecular Formula: C8H12BNO2
InChi Key: RIIPFHVHLXPMHQ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)B(O)O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)