missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D44281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D44281G Supplier TCI America Supplier No. D44281G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-(Dimethylamino)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 28611-39-4
Melting Point 227°C
Color White-Yellow
Molecular Formula C8H12BNO2
Quantity 1 g
MDL Number MFCD01074642
Synonym 4-dimethylamino phenylboronic acid, 4-dimethylamino phenyl boronic acid, 4-n,n-dimethylamino phenylboronic acid, 4-dimethylamino benzeneboronic acid, 4-dimethylaminophenylboronic acid, 4-dimethylaminophenyl boronic acid, 4-dimethylamino phenyl boranediol, 4-n,n-dimethylphenylboronic acid
InChI Key RIIPFHVHLXPMHQ-UHFFFAOYSA-N
SMILES CN(C)C1=CC=C(C=C1)B(O)O
IUPAC Name [4-(dimethylamino)phenyl]boronic acid
Molecular Weight (g/mol) 165.00
PubChem CID 2734344
Formula Weight 165.00
Percent Purity 97.0 to 113.0%
Physical Form Crystalline Powder
Show More Show Less