accessibility menu, dialog, popup

UserName

CAS RN 2856-63-5

IUPAC Name: 2-(2-chlorophenyl)acetonitrile
Synonyms: 1-chloro-2-cyanomethylbenzene
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: MRDUURPIPLIGQX-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1CC#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)