missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl Cyanide 98.0+%, TCI America™

Catalog No. C1125500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1125500G 500 g
C112525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1125500G Supplier TCI America Supplier No. C1125500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2856-63-5
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00001898
InChI Key MRDUURPIPLIGQX-UHFFFAOYSA-N
Synonym 2-chlorobenzyl cyanide, 2-chlorophenylacetonitrile, 2-2-chlorophenyl acetonitrile, o-chlorobenzyl cyanide, benzeneacetonitrile, 2-chloro, 2-chlorobenzeneacetonitrile, 2-chlorobenzylcyanide, 2-chlorophenyl acetonitrile, o-chlorophenyl acetonitrile, acetonitrile, o-chlorophenyl
PubChem CID 76112
IUPAC Name 2-(2-chlorophenyl)acetonitrile
SMILES C1=CC=C(C(=C1)CC#N)Cl

Specifications

Color White-Yellow
Quantity 500 g
UN Number 2811
Formula Weight 151.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chlorobenzyl Cyanide
EINECSNumber (3)-1784
TSCA No