accessibility menu, dialog, popup

UserName

CH3OF

CAS RN 2836-82-0

IUPAC Name: 1-(2-fluorophenyl)propan-2-one
Synonyms: 1-(2-fluorophenyl)propan-2-one
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: BANVZEUCJHUPOI-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=CC=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)