accessibility menu, dialog, popup

UserName

FFClOOOFFCl

CAS RN 2834-23-3

IUPAC Name: 2-chloro-2,2-difluoroacetyl 2-chloro-2,2-difluoroacetate
Synonyms: 2-chloro-2,2-difluoroacetyl 2-chloro-2,2-difluoroacetate
Molecular Weight (g/mol): 242.93
Molecular Formula: C4Cl2F4O3
InChi Key: VBJIFLOSOQGDRZ-UHFFFAOYSA-N
SMILES: FC(F)(Cl)C(=O)OC(=O)C(F)(F)Cl
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)