missing translation for 'onlineSavingsMsg'
Learn More

Chlorodifluoroacetic Anhydride 98.0+%, TCI America™

Catalog No. C150325G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C150325G 25 g
C150310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C150325G Supplier TCI America Supplier No. C150325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2834-23-3
Molecular Formula C4Cl2F4O3
Molecular Weight (g/mol) 242.93
MDL Number MFCD00000774
InChI Key VBJIFLOSOQGDRZ-UHFFFAOYSA-N
Synonym chlorodifluoroacetic anhydride, acetic acid, chlorodifluoro-, anhydride, acetic acid, 2-chloro-2,2-difluoro-, 1,1'-anhydride, chlorodifluoroacetic anhyride, chlorodifluoroacetic acid anhydride, 2-chloro-2,2-difluoroacetic anhydride, 2-chloro-2,2-difluoroacetyl 2-chloro-2,2-difluoroacetate
PubChem CID 76074
IUPAC Name 2-chloro-2,2-difluoroacetyl 2-chloro-2,2-difluoroacetate
SMILES FC(F)(Cl)C(=O)OC(=O)C(F)(F)Cl

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Formula Weight 242.93
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Chlorodifluoroacetic Anhydride