accessibility menu, dialog, popup

UserName

CAS RN 28315-93-7

IUPAC Name: 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 5-hydroxy-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: YPPZCRZRQHFRBH-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1CCCC2=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)