accessibility menu, dialog, popup

UserName

CAS RN 2824-57-9

NH(S)HNOCH3OOCH3

CAS RN 2824-57-9

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)