accessibility menu, dialog, popup

UserName

CAS RN 28177-48-2

IUPAC Name: 2,6-difluorophenol
Synonyms: phenol, 2,6-difluoro-
Molecular Weight (g/mol): 130.09
Molecular Formula: C6H4F2O
InChi Key: CKKOVFGIBXCEIJ-UHFFFAOYSA-N
SMILES: OC1=C(F)C=CC=C1F
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)