accessibility menu, dialog, popup

UserName

CH3OO(S)CH3HO

CAS RN 27871-49-4

IUPAC Name: methyl (2S)-2-hydroxypropanoate
Synonyms: methyl (2S)-2-hydroxypropanoate
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: LPEKGGXMPWTOCB-VKHMYHEASA-N
SMILES: COC(=O)[C@H](C)O
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (4)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)