missing translation for 'onlineSavingsMsg'
Learn More

Methyl L-(-)-Lactate 98.0+%, TCI America™

Catalog No. L0163500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0163500G 500 g
L016325G 25 g
L0163100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L0163500G Supplier TCI America Supplier No. L0163500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27871-49-4
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00064265
InChI Key LPEKGGXMPWTOCB-VKHMYHEASA-N
Synonym methyl s---lactate, methyl l-lactate, s-methyl lactate, --methyl l-lactate, s-lactic acid methyl ester, methyl 2s-2-hydroxypropanoate, --methyl lactate, methyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, methyl ester, 2s, --lactic acid methyl ester
PubChem CID 94386
ChEBI CHEBI:83222
IUPAC Name methyl (2S)-2-hydroxypropanoate
SMILES COC(=O)[C@H](C)O

Specifications

Color Colorless
Boiling Point 140°C
Quantity 500 g
UN Number 3272
Formula Weight 104.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl L-(-)-Lactate
EINECSNumber (2)-1370
TSCA Yes