accessibility menu, dialog, popup

UserName

H3COOCH3HOO

CAS RN 2785-98-0

IUPAC Name: 2,5-dimethoxybenzoic acid
Synonyms: 6-methoxy-m-anisic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: NYJBTJMNTNCTCP-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(=C1)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)