accessibility menu, dialog, popup

UserName

H3COOOCH3

CAS RN 27798-60-3

IUPAC Name: methyl 2-(2-methoxyphenyl)acetate
Synonyms: methyl 2-(2-methoxyphenyl)acetate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: BNQRSYFOIRGRKV-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC=C1OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results