missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Methoxyphenylacetate 98.0+%, TCI America™

Catalog No. M13295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13295G 5 g
M132925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13295G Supplier TCI America Supplier No. M13295G

Chemical Identifiers

CAS 27798-60-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD02093479
InChI Key BNQRSYFOIRGRKV-UHFFFAOYSA-N
Synonym 2-Methoxyphenylacetic Acid Methyl Ester
PubChem CID 99590
IUPAC Name methyl 2-(2-methoxyphenyl)acetate
SMILES COC(=O)CC1=CC=CC=C1OC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Methoxyphenylacetate