accessibility menu, dialog, popup

UserName

CAS RN 2736-37-0

IUPAC Name: 2-methylpropanoyl bromide
Synonyms: 2-methylpropanoyl bromide
Molecular Weight (g/mol): 151
Molecular Formula: C4H7BrO
InChi Key: ICNCZFQYZKPYMS-UHFFFAOYSA-N
SMILES: CC(C)C(Br)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results