missing translation for 'onlineSavingsMsg'
Learn More

Isobutyryl Bromide 98.0+%, TCI America™

Catalog No. I011425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I011425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I011425G Supplier TCI America Supplier No. I011425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2736-37-0
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151.003
MDL Number MFCD00059468
InChI Key ICNCZFQYZKPYMS-UHFFFAOYSA-N
PubChem CID 520309
IUPAC Name 2-methylpropanoyl bromide
SMILES CC(C)C(=O)Br

Specifications

Color Yellow
Boiling Point 117°C
Quantity 25 g
UN Number 2920
Formula Weight 151.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyryl Bromide