accessibility menu, dialog, popup

UserName

CAS RN 27318-90-7

IUPAC Name: 5,6-dihydro-1,10-phenanthroline-5,6-dione
Synonyms: 1,10-phenanthroline-5,6-dione
Molecular Weight (g/mol): 210.19
Molecular Formula: C12H6N2O2
InChi Key: KCALAFIVPCAXJI-UHFFFAOYSA-N
SMILES: O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)