accessibility menu, dialog, popup

UserName

CAS RN 2713-34-0

IUPAC Name: 3,5-difluorophenol
Synonyms: 3,5-difluorophenol
Molecular Weight (g/mol): 130.09
Molecular Formula: C6H4F2O
InChi Key: HJSSBIMVTMYKPD-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC(F)=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)