missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenol 98.0+%, TCI America™

Catalog No. D25695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25695G 5 g
D256925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25695G Supplier TCI America Supplier No. D25695G

Chemical Identifiers

CAS 2713-34-0
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00002255
InChI Key HJSSBIMVTMYKPD-UHFFFAOYSA-N
Synonym phenol, 3,5-difluoro, 3,5-difluoro phenol, phenol,3,5-difluoro, 3,5difluorophenol, 3,5-difluorphenol, 3.5-difluorophenol, 3,5-difluoro-phenol, phenol derivative, 6, pubchem1496, acmc-209gvc
PubChem CID 75928
IUPAC Name 3,5-difluorophenol
SMILES OC1=CC(F)=CC(F)=C1

Specifications

Melting Point 54°C
Boiling Point 68°C
Quantity 5 g
Formula Weight 130.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Difluorophenol