accessibility menu, dialog, popup

UserName

NH2O

CAS RN 2688-84-8

IUPAC Name: 2-phenoxyaniline
Synonyms: O-phenoxyaniline
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: NMFFUUFPJJOWHK-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1OC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)