accessibility menu, dialog, popup

UserName

CH3H3CNNNNNHOH3CNHOCH3

CAS RN 267402-71-1

IUPAC Name: N2,N6-bis[(4-methoxyphenyl)methyl]-9-(propan-2-yl)-9H-purine-2,6-diamine
Synonyms: 9-isopropyl-N2,N6-bis[(4-methoxyphenyl)methyl]purine-2,6-diamine
Molecular Weight (g/mol): 432.53
Molecular Formula: C24H28N6O2
InChi Key: IDCOTQWQVPRTNK-UHFFFAOYSA-N
SMILES: COC1=CC=C(CNC2=NC(NCC3=CC=C(OC)C=C3)=C3N=CN(C(C)C)C3=N2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results