missing translation for 'onlineSavingsMsg'
Learn More

Myoseverin 98.0+%, TCI America™

Catalog No. M237310MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M237310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M237310MG Supplier TCI America Supplier No. M237310MG
Only null left

Chemical Identifiers

CAS 267402-71-1
Molecular Formula C24H28N6O2
Molecular Weight (g/mol) 432.528
MDL Number MFCD02683596
InChI Key IDCOTQWQVPRTNK-UHFFFAOYSA-N
Synonym 9-Isopropyl-2,6-bis(4-methoxybenzylamino)purine
PubChem CID 4273
IUPAC Name 2-N,6-N-bis[(4-methoxyphenyl)methyl]-9-propan-2-ylpurine-2,6-diamine
SMILES CC(C)N1C=NC2=C1N=C(N=C2NCC3=CC=C(C=C3)OC)NCC4=CC=C(C=C4)OC

Specifications

Melting Point 108°C
Color Yellow
Quantity 10 mg
Formula Weight 432.53
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Myoseverin
TSCA No
Recommended Storage Freezer