CAS RN 265670-72-2
IUPAC Name:
6-fluoro-2,3-dimethoxybenzoic acid
Synonyms:
6-fluoro-2,3-dimethoxybenzoic acid
Molecular Weight (g/mol):
200.17
Molecular Formula:
C9H9FO4
InChi Key:
OXYSHPNWRGUQPS-UHFFFAOYSA-N
SMILES:
COC1=C(OC)C(C(O)=O)=C(F)C=C1
1
–
1
of
1
results
1
Promotions Available
6-Fluoro-2,3-dimethoxybenzoic acid, 95%
6-Fluoro-2,3-dimethoxybenzoic acid, 95%
CAS: 265670-72-2 Molecular Formula: C9H9FO4 Molecular Weight (g/mol): 200.165 MDL Number: MFCD13193706 InChI Key: OXYSHPNWRGUQPS-UHFFFAOYSA-N Synonym: benzoic acid, 6-fluoro-2,3-dimethoxy,5,6-dimethoxy-2-fluorobenzoic acid,6-fluoro-2,3-dimethoxybenzoicacid,2,3-dimethoxy-6-fluorobenzoic acid,#,benzoicacid, 6-fluoro-2,3-dimethoxy,benzoic acid, 2-fluoro-5,6-dimethoxy PubChem CID: 608235 IUPAC Name: 6-fluoro-2,3-dimethoxybenzoic acid SMILES: COC1=C(C(=C(C=C1)F)C(=O)O)OC