missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-2,3-dimethoxybenzoic acid, 95%

Catalog No. AAH66209MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66209MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66209MD Supplier Thermo Scientific Chemicals Supplier No. H66209MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 265670-72-2 | C9H9FO4 | 200.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 265670-72-2
Molecular Formula C9H9FO4
Molecular Weight (g/mol) 200.165
MDL Number MFCD13193706
InChI Key OXYSHPNWRGUQPS-UHFFFAOYSA-N
Synonym benzoic acid, 6-fluoro-2,3-dimethoxy, 5,6-dimethoxy-2-fluorobenzoic acid, 6-fluoro-2,3-dimethoxybenzoicacid, 2,3-dimethoxy-6-fluorobenzoic acid, #, benzoicacid, 6-fluoro-2,3-dimethoxy, benzoic acid, 2-fluoro-5,6-dimethoxy
PubChem CID 608235
IUPAC Name 6-fluoro-2,3-dimethoxybenzoic acid
SMILES COC1=C(C(=C(C=C1)F)C(=O)O)OC

Specifications

Quantity 250 mg
Beilstein 8482222
Formula Weight 200.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 6-Fluoro-2,3-dimethoxybenzoic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only