CAS RN 2627-86-3
(S)-(-)-1-Phenylethylamine, 99+%, produced by BASF AG
CAS: 2627-86-3 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.18 MDL Number: MFCD00064406 InChI Key: RQEUFEKYXDPUSK-ZETCQYMHSA-N Synonym: s---1-phenylethylamine,s-1-phenylethanamine,1s-1-phenylethanamine,s---alpha-methylbenzylamine,s-1-phenylethylamine,l--alpha-methylbenzylamine,l-alpha-methylbenzylamine,s-alpha-methylbenzenemethanamine,--alpha-phenethylamine,l---1-phenylethylamine PubChem CID: 75818 ChEBI: CHEBI:35321 IUPAC Name: (1S)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
(S)-(-)-1-Phenylethylamine, 98%
CAS: 2627-86-3 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.183 MDL Number: MFCD00064406 InChI Key: RQEUFEKYXDPUSK-ZETCQYMHSA-N Synonym: s---1-phenylethylamine,s-1-phenylethanamine,1s-1-phenylethanamine,s---alpha-methylbenzylamine,s-1-phenylethylamine,l--alpha-methylbenzylamine,l-alpha-methylbenzylamine,s-alpha-methylbenzenemethanamine,--alpha-phenethylamine,l---1-phenylethylamine PubChem CID: 75818 ChEBI: CHEBI:35321 IUPAC Name: (1S)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
L(-)-alpha-Methylbenzylamine, 99+%, (99% ee)
CAS: 2627-86-3 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.18 InChI Key: RQEUFEKYXDPUSK-ZETCQYMHSA-N Synonym: s---1-phenylethylamine,s-1-phenylethanamine,1s-1-phenylethanamine,s---alpha-methylbenzylamine,s-1-phenylethylamine,l--alpha-methylbenzylamine,l-alpha-methylbenzylamine,s-alpha-methylbenzenemethanamine,--alpha-phenethylamine,l---1-phenylethylamine PubChem CID: 75818 ChEBI: CHEBI:35321 IUPAC Name: (1S)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
(S)-(-)-1-Phenylethylamine, ChiPros 99+%, ee 99.5%
CAS: 2627-86-3 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.183 MDL Number: MFCD00064406 InChI Key: RQEUFEKYXDPUSK-ZETCQYMHSA-N Synonym: s---1-phenylethylamine,s-1-phenylethanamine,1s-1-phenylethanamine,s---alpha-methylbenzylamine,s-1-phenylethylamine,l--alpha-methylbenzylamine,l-alpha-methylbenzylamine,s-alpha-methylbenzenemethanamine,--alpha-phenethylamine,l---1-phenylethylamine PubChem CID: 75818 ChEBI: CHEBI:35321 IUPAC Name: (1S)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N
(S)-(-)-1-Phenylethylamine 98.0+%, TCI America™
CAS: 2627-86-3 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.183 MDL Number: MFCD00064406 InChI Key: RQEUFEKYXDPUSK-ZETCQYMHSA-N Synonym: s---1-phenylethylamine,s-1-phenylethanamine,1s-1-phenylethanamine,s---alpha-methylbenzylamine,s-1-phenylethylamine,l--alpha-methylbenzylamine,l-alpha-methylbenzylamine,s-alpha-methylbenzenemethanamine,--alpha-phenethylamine,l---1-phenylethylamine PubChem CID: 75818 ChEBI: CHEBI:35321 IUPAC Name: (1S)-1-phenylethanamine SMILES: CC(C1=CC=CC=C1)N