accessibility menu, dialog, popup

UserName

H3COOOH(R)O(R)OH(S)OH

CAS RN 26266-57-9

IUPAC Name: 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl hexadecanoate
Synonyms: sorbitan palmitate
Molecular Weight (g/mol): 402.57
Molecular Formula: C22H42O6
InChi Key: IYFATESGLOUGBX-CBOZIWPYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results