missing translation for 'onlineSavingsMsg'
Learn More

Span 40 (=Sorbitan Monopalmitate), TCI America™

Catalog No. S006125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S006125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S006125G Supplier TCI America Supplier No. S006125G
Only null left

Chemical Identifiers

CAS 26266-57-9
Molecular Formula C22H42O6
Molecular Weight (g/mol) 402.572
MDL Number MFCD00080946
InChI Key IYFATESGLOUGBX-NDUCAMMLSA-N
Synonym Arlacel 40, Sorbitan Monopalmitate
PubChem CID 70687305
IUPAC Name [2-[(2R,3R,4S)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(C1C(C(CO1)O)O)O

Specifications

Melting Point 47°C
Quantity 25 g
Physical Form Pellets
Chemical Name or Material Span 40 (=Sorbitan Monopalmitate)
EINECSNumber (8)-0063
RTECSNumber WG2932900
TSCA Yes