accessibility menu, dialog, popup

UserName

IO

CAS RN 26260-02-6

IUPAC Name: 2-iodobenzaldehyde
Synonyms: 2-iodobenzaldehyde
Molecular Weight (g/mol): 232.02
Molecular Formula: C7H5IO
InChi Key: WWKKTHALZAYYAI-UHFFFAOYSA-N
SMILES: IC1=CC=CC=C1C=O
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)