missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzaldehyde, 98%

Catalog No. AAL1744606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1744606 5 g
AAL1744603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1744606 Supplier Thermo Scientific Chemicals Supplier No. L1744606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26260-02-6 | C7H5IO | 232.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26260-02-6
Molecular Formula C7H5IO
Molecular Weight (g/mol) 232.02
MDL Number MFCD00039570
InChI Key WWKKTHALZAYYAI-UHFFFAOYSA-N
Synonym benzaldehyde, 2-iodo, o-iodobenzaldehyde, iodobenzaldehyde, benzaldehyde,iodo, 2-iodobenzenealdehyde, pubchem3914, acmc-209upi, 2-iodobenzaldehyde, ksc203m8l
PubChem CID 643439
IUPAC Name 2-iodobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)I

Specifications

Melting Point 36°C to 37°C
Boiling Point 102°C (3 mmHg)
Quantity 5 g
Beilstein 1927296
Sensitivity Air and light sensitive
Formula Weight 232.02
Percent Purity 98%
Chemical Name or Material 2-Iodobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only