CAS RN 26115-70-8
IUPAC Name:
tris[3-(trimethoxysilyl)propyl]-1,3,5-triazinane-2,4,6-trione
Synonyms:
tris[3-(trimethoxysilyl)propyl]-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol):
615.86
Molecular Formula:
C21H45N3O12Si3
InChi Key:
QWOVEJBDMKHZQK-UHFFFAOYSA-N
SMILES:
CO[Si](CCCN1C(=O)N(CCC[Si](OC)(OC)OC)C(=O)N(CCC[Si](OC)(OC)OC)C1=O)(OC)OC
1
–
1
of
1
results
1
Tris[3-(trimethoxysilyl)propyl] Isocyanurate 95.0+%, TCI America™
CAS: 26115-70-8 Molecular Formula: C21H45N3O12Si3 Molecular Weight (g/mol): 615.855 MDL Number: MFCD00054746 InChI Key: QWOVEJBDMKHZQK-UHFFFAOYSA-N Synonym: Isocyanuric Acid Tris[3-(trimethoxysilyl)propyl] Ester PubChem CID: 117734 IUPAC Name: 1,3,5-tris(3-trimethoxysilylpropyl)-1,3,5-triazinane-2,4,6-trione SMILES: CO[Si](CCCN1C(=O)N(C(=O)N(C1=O)CCC[Si](OC)(OC)OC)CCC[Si](OC)(OC)OC)(OC)OC