missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris[3-(trimethoxysilyl)propyl] Isocyanurate 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 236°C |
| Quantity | 5 g |
| Formula Weight | 615.86 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Tris[3-(trimethoxysilyl)propyl] Isocyanurate |
Safety and Handling
| EINECSNumber | (2)-2964 |
| RTECSNumber | XZ2025000 |
| TSCA | Yes |