accessibility menu, dialog, popup

UserName

CAS RN 26106-63-8

IUPAC Name: oxolane-2,3,4,5-tetracarboxylic acid
Synonyms: oxolane-2,3,4,5-tetracarboxylic acid
Molecular Weight (g/mol): 248.14
Molecular Formula: C8H8O9
InChi Key: UFOIOXZLTXNHQH-UHFFFAOYNA-N
SMILES: OC(=O)C1OC(C(C1C(O)=O)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results