accessibility menu, dialog, popup

UserName

ONOClClO

CAS RN 25784-91-2

IUPAC Name: 2-chloro-5-nitrobenzoyl chloride
Synonyms: 2-chloro-5-nitrobenzoyl chloride
Molecular Weight (g/mol): 220.01
Molecular Formula: C7H3Cl2NO3
InChi Key: OGLKKYALUKXVPQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C(=C1)C(Cl)=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)