missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzoyl chloride, 96%, Thermo Scientific Chemicals

Catalog No. AAL1752314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1752314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1752314 Supplier Thermo Scientific Chemicals Supplier No. L1752314

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25784-91-2
Molecular Formula C7H3Cl2NO3
Molecular Weight (g/mol) 220.005
MDL Number MFCD00059180
InChI Key OGLKKYALUKXVPQ-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzoylchloride, benzoyl chloride, 2-chloro-5-nitro, acmc-1cq6z, 2-chloro-5nitrobenzoyl chloride, 2-chloro-5-nitro-benzoylchlorid, 2-chloro-5-nitro-benzoic chloride, 2-chloro-5-nitro-benzoyl chloride, benzoyl chloride,2-chloro-5-nitro, 2-chloranyl-5-nitro-benzoyl chloride, 2-chloro-5-nitrobenzoic acid chloride
PubChem CID 117637
IUPAC Name 2-chloro-5-nitrobenzoyl chloride
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)Cl)Cl

Specifications

Melting Point 58°C to 60°C
Boiling Point 157°C to 158°C (11 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 2213721
Sensitivity Moisture sensitive
Formula Weight 220.01
Percent Purity 96%
Chemical Name or Material 2-Chloro-5-nitrobenzoyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 247-262-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only