missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzoyl Chloride 98.0+%, TCI America™

Catalog No. C049925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C049925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C049925G Supplier TCI America Supplier No. C049925G

Chemical Identifiers

CAS 25784-91-2
Molecular Formula C7H3Cl2NO3
Molecular Weight (g/mol) 220.005
MDL Number MFCD00059180
InChI Key OGLKKYALUKXVPQ-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzoylchloride, benzoyl chloride, 2-chloro-5-nitro, acmc-1cq6z, 2-chloro-5nitrobenzoyl chloride, 2-chloro-5-nitro-benzoylchlorid, 2-chloro-5-nitro-benzoic chloride, 2-chloro-5-nitro-benzoyl chloride, benzoyl chloride,2-chloro-5-nitro, 2-chloranyl-5-nitro-benzoyl chloride, 2-chloro-5-nitrobenzoic acid chloride
PubChem CID 117637
IUPAC Name 2-chloro-5-nitrobenzoyl chloride
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)Cl)Cl

Specifications

Melting Point 58°C
Boiling Point 158°C
Quantity 25 g
UN Number 3261
Formula Weight 220.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrobenzoyl Chloride